Gene Transcripts

Gene Symbol
Crygc
Entrez Gene ID
24277
Gene Name
crystallin, gamma C
Gene Aliases
Cryg, Cryg3, Len
Chromosome Location
Chr. 9: 71786246 - 71788281 on Build Rnor_6.0
UniGene ID
Rn.81172
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081660.1 NP_001075129.1 3 302
NM_033483.1 NP_264077.1 2 285
GenBank mRNA BF523642.1 - 2 291
J00717.1 - 2 264

View Full Product Details