Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022506.3 | NP_071951.1 | 4 | 286 | |
GenBank mRNA | BC062228.1 | - | 439 | ||
FQ211300.1 | - | 4 | 304 | ||
FQ216848.1 | - | 4 | 285 | ||
FQ221256.1 | - | 4 | 285 | ||
FQ221939.1 | - | 4 | 284 | ||
FQ222938.1 | - | 4 | 289 | ||
FQ222963.1 | - | 4 | 285 | ||
FQ223725.1 | - | 4 | 284 | ||
FQ226734.1 | - | 284 | |||
FQ228354.1 | - | 4 | 285 | ||
FQ228461.1 | - | 4 | 285 | ||
FQ228715.1 | - | 4 | 282 | ||
X04809.1 | - | 4 | 286 |