Gene Transcripts

Gene Symbol
Vom2r35
Entrez Gene ID
292615
Gene Name
vomeronasal 2 receptor, 35
Gene Aliases
RGD1559147
Chromosome Location
Chr. 1: 66004349 - 66021670 on Build Rnor_6.0
UniGene ID
Rn.217939
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099473.1 NP_001092943.1 2 982

View Full Product Details