Gene Transcripts
- Gene Symbol
- DDAH2
- Entrez Gene ID
- 23564
- Gene Name
- dimethylarginine dimethylaminohydrolase 2
- Gene Aliases
- DDAH, DDAHII, G6a, HEL-S-277, NG30
- Chromosome Location
- Chr. 6: 31727037 - 31730265 on Build GRCh38
- UniGene ID
- Hs.247362
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303007.1 |
NP_001289936.1
|
2, 3 |
1508 |
|
NM_001303008.1 |
NP_001289937.1
|
3, 4 |
608 |
|
NM_013974.2 |
NP_039268.1
|
3, 4 |
683 |
|
XM_011514448.2 |
XP_011512750.1
|
3, 4 |
759 |
|
GenBank mRNA |
AF070667.1 |
- |
3, 4 |
680 |
|
AF087894.1 |
- |
3, 4 |
680 |
|
AK026191.1 |
- |
2, 3 |
1035 |
|
AK098671.1 |
- |
3, 4 |
670 |
|
BC001435.2 |
- |
3, 4 |
595 |
3138832 |
EB387244.1 |
- |
3, 4 |
582 |
|
FJ224290.1 |
- |
3, 4 |
680 |
|
View Full Product Details