Gene Transcripts

Gene Symbol
CARHSP1
Entrez Gene ID
23589
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
CRHSP-24, CRHSP24, CSDC1
Chromosome Location
Chr. 16: 8852942 - 8869012 on Build GRCh38
UniGene ID
Hs.632184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042476.2 NP_001035941.1 4 1036
NM_001278260.1 NP_001265189.1 5 844
NM_001278261.1 NP_001265190.1 5 983
NM_001278262.1 NP_001265191.1 5 942
NM_001278263.1 NP_001265192.1 4 718
NM_001278264.1 NP_001265193.1 4 860
NM_001278265.1 NP_001265194.1 4 852
NM_001278266.1 NP_001265195.1 4 828
NM_014316.3 NP_055131.2 4 730
XM_005255228.4 XP_005255285.3 4 894
XM_005255229.4 XP_005255286.1 4 706
XM_011522444.2 XP_011520746.1 4 705
GenBank mRNA AF115345.1 - 3 686
BC001333.1 - 4 1036
BC003366.1 - 4 727 2900952
BC108283.1 - 4 717 4430943

View Full Product Details