Gene Transcripts

Gene Symbol
CARHSP1
Entrez Gene ID
23589
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
CRHSP-24, CRHSP24, CSDC1
Chromosome Location
Chr. 16: 8852942 - 8869012 on Build GRCh38
UniGene ID
Hs.632184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042476.2 NP_001035941.1 3, 4 638
NM_001278260.1 NP_001265189.1 4, 5 446
NM_001278261.1 NP_001265190.1 4, 5 585
NM_001278262.1 NP_001265191.1 4, 5 544
NM_001278263.1 NP_001265192.1 3, 4 320
NM_001278264.1 NP_001265193.1 3, 4 462
NM_001278265.1 NP_001265194.1 3, 4 454
NM_001278266.1 NP_001265195.1 3, 4 430
NM_014316.3 NP_055131.2 3, 4 332
XM_005255228.4 XP_005255285.3 3, 4 496
XM_005255229.4 XP_005255286.1 3, 4 308
XM_011522444.2 XP_011520746.1 3, 4 307
GenBank mRNA AF115345.1 - 2, 3 289
AK311777.1 - 3, 4 326
BC001333.1 - 3, 4 638
BC003366.1 - 3, 4 329 2900952
BC108283.1 - 3, 4 312 4430943
BM693675.1 - 3, 4 462
BM920489.1 - 3, 4 320
CA396216.1 - 3, 4 389
DA736976.1 - 4, 5 585
DB101255.1 - 4, 5 440
HY005510.1 - 3, 4 455
HY090855.1 - 3, 4 332

View Full Product Details