Gene Transcripts

Gene Symbol
RAB38
Entrez Gene ID
23682
Gene Name
RAB38, member RAS oncogene family
Gene Aliases
NY-MEL-1, rrGTPbp
Chromosome Location
Chr. 11: 87809490 - 88175505 on Build GRCh38
UniGene ID
Hs.591975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022337.2 NP_071732.1 3 600
GenBank mRNA AF136172.1 - 3 521
AF235022.1 - 3 564
BC015808.1 - 3 566 3916084
BT009842.1 - 3 517

View Full Product Details