Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001755.2 | NP_001746.1 | 4 | 602 | |
NM_022845.2 | NP_074036.1 | 4 | 602 | ||
XM_006721321.2 | XP_006721384.1 | 3 | 447 | ||
XM_017023812.1 | XP_016879301.1 | 3 | 447 | ||
GenBank mRNA | AA020956.1 | - | 4 | 407 | |
AF294326.1 | - | 4 | 341 | ||
AK290462.1 | - | 4 | 360 | ||
AK291834.1 | - | 4 | 344 | ||
BC018509.1 | - | 4 | 602 | 3867061 | |
BT006862.1 | - | 4 | 341 | ||
L20298.1 | - | 4 | 320 |