Gene Transcripts

Gene Symbol
PHF19
Entrez Gene ID
26147
Gene Name
PHD finger protein 19
Gene Aliases
MTF2L1, PCL3, TDRD19B
Chromosome Location
Chr. 9: 120855651 - 120901967 on Build GRCh38
UniGene ID
Hs.460124
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009936.2 NP_001009936.1 3 470
NM_001286840.1 NP_001273769.1 3 341
NM_001286843.1 NP_001273772.1 3 470
NM_015651.2 NP_056466.1 3 470
XM_005251906.2 XP_005251963.1 3 278
XM_011518509.2 XP_011516811.1 3 321
XM_011518511.2 XP_011516813.1 3 804
XM_011518515.2 XP_011516817.1 3 805
XM_011518516.2 XP_011516818.1 3 807
XM_017014612.1 XP_016870101.1 3 310
XM_017014613.1 XP_016870102.1 3 808
GenBank mRNA AK122744.1 - 3 330
BC022374.1 - 3 330
BC108663.1 - 3 437
BC125076.1 - 2 242 40117657
BC125077.1 - 2 242 40117658
BX640713.1 - 3 342

View Full Product Details