Gene Transcripts

Gene Symbol
TOR2A
Entrez Gene ID
27433
Gene Name
torsin family 2 member A
Gene Aliases
TORP1
Chromosome Location
Chr. 9: 127731524 - 127735387 on Build GRCh38
UniGene ID
Hs.444106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085347.2 NP_001078816.1 3 642
NM_001134430.2 NP_001127902.1 3 642
NM_001134431.2 NP_001127903.1 2 376
NM_001252018.1 NP_001238947.1 2 376
NM_001252021.1 NP_001238950.1 2 373
NM_001252023.1 NP_001238952.1 2 376
NM_130459.3 NP_569726.2 3 642
XM_011518554.1 XP_011516856.1 3 677
GenBank mRNA AK027677.1 - 3 577
AK055034.1 - 2 321
AK075520.1 - 3 585
AK092388.1 - 1 1893
AY358180.1 - 3 609
BC062340.1 - 2 341
BC080527.1 - 2 302
BC100907.1 - 2 311
BC100908.2 - 2 311
BC100909.2 - 2 306
BC100910.1 - 2 311
BF115698.1 - 2 172
BM695589.1 - 2 317
BM772287.1 - 2 319
CN263138.1 - 3 644
DB168410.1 - 1 54

View Full Product Details