Gene Transcripts

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
Chromosome Location
Chr. 4: 25377226 - 25418498 on Build GRCh38
UniGene ID
Hs.152173
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286756.1 NP_001273685.1 20 1518
NM_013367.2 NP_037499.2 20 1515
XM_005248159.2 XP_005248216.1 7 412
XM_011513838.1 XP_011512140.1 20 1395
XM_011513839.1 XP_011512141.1 20 1439
XM_017008138.1 XP_016863627.1 20 1444
GenBank mRNA AF191338.1 - 19 1386
AK023597.1 - 17 1129
AK292336.1 - 20 1435
AL353932.1 - 3 88
BC059383.1 - 20 1488 30349524

View Full Product Details