Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_037144.1 | 3 | 1496 | ||
GenBank mRNA | AF103731.1 | - | 3 | 1496 | |
AF380162.1 | - | 1 | 1325 | ||
AK024218.1 | - | 1 | 471 | ||
BC028914.1 | - | 1 | 1337 | ||
BC063575.1 | - | 3 | 1377 | 3881751 |