Gene Transcripts
- Gene Symbol
- RTFDC1
- Entrez Gene ID
- 51507
- Gene Name
- replication termination factor 2 domain containing 1
- Gene Aliases
- C20orf43, CDAO5, HSPC164, SHUJUN-3
- Chromosome Location
- Chr. 20: 56468585 - 56518886 on Build GRCh38
- UniGene ID
- Hs.517134
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001283036.1 |
NP_001269965.1
|
1, 2 |
181 |
|
NM_001283037.1 |
NP_001269966.1
|
1, 2 |
181 |
|
NM_016407.4 |
NP_057491.2
|
1, 2 |
181 |
|
GenBank mRNA |
AF117231.1 |
- |
1, 2 |
129 |
|
AF161513.1 |
- |
1, 2 |
138 |
|
AF161518.1 |
- |
1, 2 |
107 |
|
AF212244.1 |
- |
1, 2 |
118 |
|
AK000481.1 |
- |
1, 2 |
111 |
|
AK026118.1 |
- |
1, 2 |
109 |
|
AK225135.1 |
- |
1, 2 |
99 |
|
AK293662.1 |
- |
1, 2 |
109 |
|
BC003359.1 |
- |
1, 2 |
108 |
2900332 |
BG468509.1 |
- |
1, 2 |
138 |
|
BP307634.1 |
- |
1, 2 |
175 |
|
BU537939.1 |
- |
1, 2 |
91 |
|
CR983692.1 |
- |
1, 2 |
92 |
|
View Full Product Details