Gene Transcripts
- Gene Symbol
- CDKN3
- Entrez Gene ID
- 1033
- Gene Name
- cyclin dependent kinase inhibitor 3
- Gene Aliases
- CDI1, CIP2, KAP, KAP1
- Chromosome Location
- Chr. 14: 54396939 - 54420218 on Build GRCh38
- UniGene ID
- Hs.84113
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130851.1 |
NP_001124323.1
|
6, 7 |
547 |
|
NM_005192.3 |
NP_005183.2
|
7, 8 |
667 |
|
GenBank mRNA |
AF213033.1 |
- |
6, 7 |
553 |
|
AF213036.1 |
- |
6, 7 |
553 |
|
AF213038.1 |
- |
5, 6 |
433 |
|
AF213039.1 |
- |
6, 7 |
553 |
|
AF213041.1 |
- |
6, 7 |
553 |
|
AF213042.1 |
- |
5, 6 |
433 |
|
AF213044.1 |
- |
4, 5 |
415 |
|
AF213045.1 |
- |
6, 7 |
535 |
|
AF213047.1 |
- |
5, 6 |
433 |
|
AF213048.1 |
- |
4, 5 |
415 |
|
AF213053.1 |
- |
6, 7 |
553 |
|
AK098653.1 |
- |
7, 8 |
667 |
|
AY257474.1 |
- |
6, 7 |
553 |
|
AY257475.1 |
- |
3, 4 |
253 |
|
BC064965.1 |
- |
6, 7 |
507 |
6143626 |
BT019451.1 |
- |
6, 7 |
553 |
|
CR407666.1 |
- |
6, 7 |
553 |
|
EF560750.1 |
- |
7, 8 |
593 |
|
KP966095.1 |
- |
6, 7 |
584 |
|
L25876.1 |
- |
7, 8 |
614 |
|
L27711.1 |
- |
7, 8 |
604 |
|
U02681.1 |
- |
7, 8 |
584 |
|
View Full Product Details