Gene Transcripts

Gene Symbol
UGT1A10
Entrez Gene ID
54575
Gene Name
UDP glucuronosyltransferase family 1 member A10
Gene Aliases
UDPGT, UGT-1J, UGT1-10, UGT1.10, UGT1J
Chromosome Location
Chr. 2: 233636477 - 233773305 on Build GRCh38
UniGene ID
Hs.554822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019075.2 NP_061948.1 1 425
GenBank mRNA AK025403.1 - 1 32
AY435137.1 - 1 379
BC020971.1 - 1 448 3850654
BC053576.1 - 1 400 4452440
BC069210.1 - 1 409 4660540
U89508.1 - 1 379

Gene Symbol
UGT1A7
Entrez Gene ID
54577
Gene Name
UDP glucuronosyltransferase family 1 member A7
Gene Aliases
UDPGT, UDPGT 1-7, UGT-1G, UGT1-07, UGT1.7, UGT1G
Chromosome Location
Chr. 2: 233681938 - 233773299 on Build GRCh38
UniGene ID
Hs.554822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019077.2 NP_061950.2 1 379
GenBank mRNA AY435142.1 - 1 379
U89507.1 - 1 379

Gene Symbol
UGT1A8
Entrez Gene ID
54576
Gene Name
UDP glucuronosyltransferase family 1 member A8
Gene Aliases
UDPGT, UDPGT 1-8, UGT-1H, UGT1-08, UGT1.8, UGT1A8S, UGT1H
Chromosome Location
Chr. 2: 233617645 - 233773299 on Build GRCh38
UniGene ID
Hs.554822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019076.4 NP_061949.3 1 442
GenBank mRNA AF030310.1 - 1 379
AF462267.1 - 1 412
AF462268.1 - 1 442
AK313488.1 - 1 425
AY435143.1 - 1 379
DQ364251.1 - 1 34

View Full Product Details