Gene Transcripts

Gene Symbol
AFTPH
Entrez Gene ID
54812
Gene Name
aftiphilin
Gene Aliases
Nbla10388
Chromosome Location
Chr. 2: 64524305 - 64593005 on Build GRCh38
UniGene ID
Hs.655167
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002243.2 NP_001002243.1 2 1192
NM_017657.4 NP_060127.3 2 1192
NM_203437.3 NP_982261.2 2 1192
XM_005264380.1 XP_005264437.1 2 1192
GenBank mRNA AB073356.1 - 1 634
AK000087.1 - 2 1167
AK222821.1 - 2 1115
AL833962.2 - 2 970
AY367088.1 - 1 852
BC047529.1 - 2 1166 5311682

View Full Product Details