Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001818.3 | NP_001809.3 | 9 | 977 | |
GenBank mRNA | AB031085.1 | - | 9 | 944 | |
AB045829.1 | - | 9 | 944 | ||
BC020744.1 | - | 9 | 977 | 4734943 | |
D26125.1 | - | 9 | 940 | ||
M33375.1 | - | 9 | 942 | ||
S68287.1 | - | 9 | 951 |