Gene Transcripts

Gene Symbol
SLC39A4
Entrez Gene ID
55630
Gene Name
solute carrier family 39 member 4
Gene Aliases
AEZ, AWMS2, ZIP4
Chromosome Location
Chr. 8: 144412414 - 144416895 on Build GRCh38
UniGene ID
Hs.521934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001280557.1 NP_001267486.1 4 436
NM_017767.2 NP_060237.2 11 2081
NM_130849.3 NP_570901.2 12 1937
XM_006716599.1 XP_006716662.1 11 1763
GenBank mRNA AK000334.1 - 11 2059
AK000489.1 - 4 431
AK000695.1 - 11 1996
AK025537.1 - 12 1931
AK056900.1 - 3 522
AK290563.1 - 12 1885
BC001688.2 - 4 408
BC062625.1 - 12 1917 5203858
KU178707.1 - 12 1831
KU178708.1 - 11 1549

View Full Product Details