Gene Transcripts

Gene Symbol
ARHGEF40
Entrez Gene ID
55701
Gene Name
Rho guanine nucleotide exchange factor 40
Gene Aliases
SOLO
Chromosome Location
Chr. 14: 21061276 - 21090248 on Build GRCh38
UniGene ID
Hs.35125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278529.1 NP_001265458.1 3 1263
NM_001278530.1 NP_001265459.1 3 1263
NM_018071.4 NP_060541.3 3 1263
XM_005267844.2 XP_005267901.1 3 1299
XM_011536937.2 XP_011535239.1 3 1299
XM_011536938.2 XP_011535240.1 3 1301
XM_017021434.1 XP_016876923.1 4 1556
XM_017021435.1 XP_016876924.1 3 1300
GenBank mRNA AK024463.1 - 1 501
AK074057.1 - 3 1208
BC142692.1 - 3 1206 40148708
BC151212.1 - 3 1206
BX248768.1 - 3 1250

View Full Product Details