Gene Transcripts

Gene Symbol
RNF20
Entrez Gene ID
56254
Gene Name
ring finger protein 20
Gene Aliases
BRE1, BRE1A, hBRE1
Chromosome Location
Chr. 9: 101533849 - 101563344 on Build GRCh38
UniGene ID
Hs.729085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019592.6 NP_062538.5 7 958
XM_011518862.1 XP_011517164.1 7 1235
GenBank mRNA AK000389.1 - 6 849
AK000697.1 - 6 849
AK022300.1 - 2 230
AK022532.1 - 7 954
AK314401.1 - 7 913
BC016691.1 - 8 913
BC032618.1 - 3 281
BC063115.1 - 7 939
BC110584.1 - 7 951 40009044
BC110585.1 - 7 951 40009047
BC152309.1 - 7 951 40009034

View Full Product Details