Gene Transcripts
- Gene Symbol
- ZNF410
- Entrez Gene ID
- 57862
- Gene Name
- zinc finger protein 410
- Gene Aliases
- APA-1, APA1
- Chromosome Location
- Chr. 14: 73886615 - 73932288 on Build GRCh38
- UniGene ID
- Hs.270869
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001242926.1 |
NP_001229855.1
|
10, 11 |
1704 |
|
NM_001242927.1 |
NP_001229856.1
|
10, 11 |
1626 |
|
NM_021188.2 |
NP_067011.1
|
11, 12 |
1845 |
|
NR_040251.1 |
|
11, 12 |
1813 |
|
GenBank mRNA |
AK223261.1 |
- |
11, 12 |
1601 |
|
AK298307.1 |
- |
12, 13 |
1568 |
|
AK298520.1 |
- |
11, 12 |
1621 |
|
AK299138.1 |
- |
10, 11 |
1624 |
|
AK303834.1 |
- |
10, 11 |
1305 |
|
AK304609.1 |
- |
11, 12 |
1359 |
|
AK315206.1 |
- |
11, 12 |
1603 |
|
BC050683.1 |
- |
10, 11 |
1514 |
5752452 |
U90919.1 |
- |
11, 12 |
1577 |
|
View Full Product Details