Gene Transcripts

Gene Symbol
NEUROD4
Entrez Gene ID
58158
Gene Name
neuronal differentiation 4
Gene Aliases
ATH-3, ATH3, Atoh3, MATH-3, MATH3, bHLHa4
Chromosome Location
Chr. 12: 55019945 - 55030017 on Build GRCh38
UniGene ID
Hs.591024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021191.2 NP_067014.2 2 501
GenBank mRNA AK314136.1 - 2 416
DA417566.1 - 2 416

View Full Product Details