Gene Transcripts

Gene Symbol
AASDHPPT
Entrez Gene ID
60496
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Gene Aliases
AASD-PPT, ACPS, CGI-80, LYS2, LYS5
Chromosome Location
Chr. 11: 106077565 - 106098692 on Build GRCh38
UniGene ID
Hs.524009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015423.2 NP_056238.2 5 892
GenBank mRNA AF136978.1 - 5 914
AF151057.1 - 5 912
AF151838.1 - 5 912
AF201943.1 - 5 861
AK293362.1 - 4 653
AK312529.1 - 5 796
BC015470.1 - 5 783 3914010
BC016728.1 - 5 812 4081713
BX537665.1 - 1 3170

View Full Product Details