Gene Transcripts

Gene Symbol
STRA6
Entrez Gene ID
64220
Gene Name
stimulated by retinoic acid 6
Gene Aliases
MCOPCB8, MCOPS9, PP14296
Chromosome Location
Chr. 15: 74179466 - 74212267 on Build GRCh38
UniGene ID
Hs.24553
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142617.1 NP_001136089.1 9 986
NM_001142618.1 NP_001136090.1 9 968
NM_001142619.1 NP_001136091.1 9 959
NM_001199040.1 NP_001185969.1 9 1102
NM_001199041.1 NP_001185970.1 9 1113
NM_001199042.1 NP_001185971.1 9 1217
NM_022369.3 NP_071764.3 9 922
XM_011521883.1 XP_011520185.1 9 888
XM_011521884.1 XP_011520186.1 9 1029
XM_011521885.2 XP_011520187.1 9 863
XM_017022478.1 XP_016877967.1 9 837
XM_017022479.1 XP_016877968.1 9 928
XM_017022480.1 XP_016877969.1 9 952
GenBank mRNA AF352728.1 - 9 788
AF352729.1 - 9 898
AF370419.2 - 9 894
AK022603.1 - 5 472
AK056125.1 - 9 983
AK091152.1 - 9 1012
AK092227.1 - 4 449
AK291966.1 - 9 858
AK298921.1 - 9 857
AK298928.1 - 7 526
AK299191.1 - 9 1102
AK302932.1 - 9 1113
AK309676.1 - 9 1197
AK310878.1 - 9 845
AY358748.1 - 9 898
AY359089.1 - 9 788
BC025256.1 - 9 894 4892821
BX537413.1 - 9 864

View Full Product Details