Gene Transcripts

Gene Symbol
C9orf16
Entrez Gene ID
79095
Gene Name
chromosome 9 open reading frame 16
Gene Aliases
EST00098
Chromosome Location
Chr. 9: 128159425 - 128163928 on Build GRCh38
UniGene ID
Hs.522412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024112.3 NP_077017.1 2 261
XM_011519004.1 XP_011517306.1 2 209
GenBank mRNA AK022885.1 - 1 2410
AK315753.1 - 2 188
BC001857.2 - 2 165 3529116
BC008887.2 - 2 165 3027807
DB493059.1 - 2 327

View Full Product Details