Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146056.2 | NP_001139528.1 | 5 | 1335 | |
NM_001146082.2 | NP_001139554.1 | 7 | 1541 | ||
NM_001146083.2 | NP_001139555.1 | 6 | 1414 | ||
NM_024298.4 | NP_077274.3 | 7 | 1541 | ||
XM_011527299.2 | XP_011525601.1 | 7 | 1745 | ||
XM_011527300.2 | XP_011525602.1 | 7 | 1136 | ||
XM_017027296.1 | XP_016882785.1 | 7 | 1709 | ||
GenBank mRNA | AK055908.1 | - | 7 | 1474 | |
AK122789.1 | - | 8 | 1259 | ||
AK123290.1 | - | 1 | 1988 | ||
AK293199.1 | - | 5 | 945 | ||
AK298689.1 | - | 5 | 1021 | ||
AK311493.1 | - | 6 | 817 | ||
BC002512.2 | - | 6 | 991 | ||
BC003164.1 | - | 7 | 1072 | 2958347 | |
BC006309.2 | - | 7 | 1331 | 4110925 | |
BC015857.1 | - | 6 | 978 | 4671596 | |
EU016381.1 | - | 6 | 993 | ||
S82470.1 | - | 5 | 847 |