Gene Transcripts
- Gene Symbol
- BEND5
- Entrez Gene ID
- 79656
- Gene Name
- BEN domain containing 5
- Gene Aliases
- C1orf165
- Chromosome Location
- Chr. 1: 48727523 - 48778603 on Build GRCh38
- UniGene ID
- Hs.475348
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001302082.1 |
NP_001289011.1
|
5, 6 |
1325 |
|
NM_024603.3 |
NP_078879.2
|
5, 6 |
1243 |
|
XM_011542141.2 |
XP_011540443.1
|
5, 6 |
3743 |
|
XM_011542142.2 |
XP_011540444.1
|
4, 5 |
1191 |
|
XM_017002331.1 |
XP_016857820.1
|
6, 7 |
1276 |
|
XM_017002332.1 |
XP_016857821.1
|
6, 7 |
1153 |
|
XM_017002334.1 |
XP_016857823.1
|
6, 7 |
1372 |
|
GenBank mRNA |
AK021650.1 |
- |
5, 6 |
1187 |
|
AK055449.1 |
- |
3, 4 |
1876 |
|
BC007932.2 |
- |
5, 6 |
1195 |
4121760 |
BC011804.2 |
- |
5, 6 |
1009 |
4336159 |
KU178776.1 |
- |
5, 6 |
1105 |
|
KU178777.1 |
- |
5, 6 |
1187 |
|
KU178778.1 |
- |
4, 5 |
971 |
|
View Full Product Details