Gene Transcripts

Gene Symbol
LRRC31
Entrez Gene ID
79782
Gene Name
leucine rich repeat containing 31
Gene Aliases
HEL-S-293
Chromosome Location
Chr. 3: 169839175 - 169869935 on Build GRCh38
UniGene ID
Hs.411295
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277127.1 NP_001264056.1 1 273
NM_001277128.1 NP_001264057.1 1 273
NM_024727.3 NP_079003.2 1 273
XM_011513158.2 XP_011511460.1 1 273
XM_011513159.2 XP_011511461.1 1 273
XM_011513160.2 XP_011511462.1 1 273
GenBank mRNA AK026912.1 - 1 269
AK097338.1 - 1 222
AY358138.1 - 1 203
BC117401.1 - 1 171 40125952
BC143569.1 - 1 172 9052080
BC143582.1 - 1 171
DA425899.1 - 1 273
DA967755.1 - 1 222
EU668335.1 - 1 222

View Full Product Details