Gene Transcripts

Gene Symbol
C10orf11
Entrez Gene ID
83938
Gene Name
chromosome 10 open reading frame 11
Gene Aliases
CDA017
Chromosome Location
Chr. 10: 75431646 - 76559220 on Build GRCh38
UniGene ID
Hs.118161
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305581.1 NP_001292510.1 3 304
NM_032024.4 NP_114413.1 2 356
NR_131178.1 4 579
XM_011540256.2 XP_011538558.1 3 298
XM_011540257.2 XP_011538559.1 3 298
XM_017016759.1 XP_016872248.1 3 298
XM_017016760.1 XP_016872249.1 3 298
XM_017016761.1 XP_016872250.1 3 298
GenBank mRNA AF267860.1 - 2 357
BM720401.1 - 2 304
BX105111.1 - 2 197
HY006640.1 - 3 295

View Full Product Details