Gene Transcripts

Gene Symbol
MIR22HG
Entrez Gene ID
84981
Gene Name
MIR22 host gene
Gene Aliases
C17orf91
Chromosome Location
Chr. 17: 1711504 - 1716272 on Build GRCh38
UniGene ID
Hs.597755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028502.1 2 402
NR_028503.1 2 402
NR_028504.1 3 485
NR_028505.1 2 402
GenBank mRNA BC119720.1 - 3 393
BC119721.1 - 3 393 40118088
BM755383.1 - 2 341
BX509423.1 - 2 339
BX640650.1 - 3 423
BX648321.1 - 2 339

View Full Product Details