Gene Transcripts
- Gene Symbol
- SYNE1
- Entrez Gene ID
- 23345
- Gene Name
- spectrin repeat containing nuclear envelope protein 1
- Gene Aliases
- 8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
- Chromosome Location
- Chr. 6: 152121684 - 152637399 on Build GRCh38
- UniGene ID
- Hs.12967
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123135.1 |
- |
|
1663 |
|
- Gene Symbol
- FOXH1
- Entrez Gene ID
- 8928
- Gene Name
- forkhead box H1
- Gene Aliases
- FAST-1, FAST1
- Chromosome Location
- Chr. 8: 144473732 - 144476335 on Build GRCh38
- UniGene ID
- Hs.708365
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BE550635.1 |
- |
1 |
89 |
|
- Gene Symbol
- KIFC2
- Entrez Gene ID
- 90990
- Gene Name
- kinesin family member C2
- Gene Aliases
- Chromosome Location
- Chr. 8: 144466246 - 144474116 on Build GRCh38
- UniGene ID
- Hs.528713
- Species
- Homo sapiens
View Full Product Details