Gene Transcripts

Gene Symbol
LRRC75A-AS1
Entrez Gene ID
125144
Gene Name
LRRC75A antisense RNA 1
Gene Aliases
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
Chromosome Location
Chr. 17: 16438987 - 16470648 on Build GRCh38
UniGene ID
Hs.368934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027160.1 2, 3 556
NR_027161.1 2, 3 609
NR_027162.1 2, 3 609
NR_027163.1 3, 4 343
NR_027164.1 3, 4 290
NR_027165.1 3, 4 211
NR_027167.1 3, 4 290
NR_027168.2 3, 4 259
NR_027169.1 3, 4 343
NR_027170.1 3, 4 211
NR_027667.1 2, 3 477
NR_045026.1 3, 4 343
NR_045028.1 2, 3 477
GenBank mRNA AK307982.1 - 2, 3 559
AK309961.1 - 3, 4 241
BC027986.1 - 2, 3 427 4940349
BC040159.1 - 2, 3 406 5539041
BI333700.1 - 2, 3 238
BP353162.1 - 3, 4 293
BP870476.1 - 3, 4 161
BQ954204.1 - 2, 3 533
BU506593.1 - 274
BU599615.1 - 3, 4 297
BX538291.1 - 3, 4 162
CB150672.1 - 3, 4 240
CR989654.1 - 2, 3 220
DA867958.1 - 3, 4 295

View Full Product Details