Gene Transcripts

Gene Symbol
FAM210A
Entrez Gene ID
125228
Gene Name
family with sequence similarity 210 member A
Gene Aliases
C18orf19, HsT2329
Chromosome Location
Chr. 18: 13663347 - 13726592 on Build GRCh38
UniGene ID
Hs.13034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098801.1 NP_001092271.1 4 1008
NM_152352.3 NP_689565.2 3 868
GenBank mRNA AK055618.1 - 4 964
BU189118.1 - 2 455

View Full Product Details