Gene Transcripts

Gene Symbol
HMGB4
Entrez Gene ID
127540
Gene Name
high mobility group box 4
Gene Aliases
dJ1007G16.5
Chromosome Location
Chr. 1: 33860475 - 33864791 on Build GRCh38
UniGene ID
Hs.568628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145205.5 NP_660206.2 2 2168
NR_033264.1 2 453
GenBank mRNA AK097707.1 - 2 2169
AK130859.1 - 1 587
AK310465.1 - 2 453
AK311985.1 - 1 554
BC021180.2 - 1 538 5266418
BC070148.1 - 2 453 6617009
BI827031.1 - 1 540
DQ207367.1 - 1 538

View Full Product Details