Gene Transcripts
- Gene Symbol
- ANKRD54
- Entrez Gene ID
- 129138
- Gene Name
- ankyrin repeat domain 54
- Gene Aliases
- LIAR
- Chromosome Location
- Chr. 22: 37830854 - 37848994 on Build GRCh38
- UniGene ID
- Hs.135259
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138797.2 |
NP_620152.1
|
1, 2 |
437 |
|
NR_036556.1 |
|
1, 2 |
437 |
|
XM_006724137.1 |
XP_006724200.1
|
1, 2 |
306 |
|
XM_011529877.2 |
XP_011528179.1
|
1, 2 |
728 |
|
XM_011529883.2 |
XP_011528185.1
|
1, 2 |
460 |
|
XM_017028593.1 |
XP_016884082.1
|
1, 2 |
707 |
|
GenBank mRNA |
BC014641.1 |
- |
1, 2 |
387 |
|
BC066909.1 |
- |
1, 2 |
404 |
4795922 |
BX401122.2 |
- |
1, 2 |
435 |
|
CD300696.1 |
- |
|
336 |
|
CR456471.1 |
- |
1, 2 |
397 |
|
View Full Product Details