Gene Transcripts

Gene Symbol
M1AP
Entrez Gene ID
130951
Gene Name
meiosis 1 associated protein
Gene Aliases
C2orf65, D6Mm5e, SPATA37
Chromosome Location
Chr. 2: 74557640 - 74648338 on Build GRCh38
UniGene ID
Hs.348645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281295.1 NP_001268224.1 5 752
NM_001281296.1 NP_001268225.1 5 752
NM_001321739.1 NP_001308668.1 5 752
NM_138804.4 NP_620159.2 5 743
XM_006711946.3 XP_006712009.1 5 1088
XM_011532548.2 XP_011530850.1 5 1089
XM_011532549.2 XP_011530851.1 5 1090
XM_011532550.2 XP_011530852.1 5 699
XM_011532551.2 XP_011530853.1 5 698
XM_011532553.2 XP_011530855.1 5 1090
GenBank mRNA AK300188.1 - 5 744
BC014602.2 - 5 721 4821635
BC025997.1 - 5 746 4823281
DR158220.1 - 3 397

View Full Product Details