Gene Transcripts

Gene Symbol
ASB8
Entrez Gene ID
140461
Gene Name
ankyrin repeat and SOCS box containing 8
Gene Aliases
PP14212
Chromosome Location
Chr. 12: 48147788 - 48157595 on Build GRCh38
UniGene ID
Hs.432699
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319296.1 NP_001306225.1 5 755
NM_001319297.1 NP_001306226.1 4 615
NM_001319298.1 NP_001306227.1 5 720
NM_001319299.1 NP_001306228.1 5 716
NM_001319300.1 NP_001306229.1 5 673
NM_001319301.1 NP_001306230.1 5 669
NM_024095.4 NP_077000.1 4 619
GenBank mRNA AF464877.1 - 4 511
AK024908.1 - 4 520
AK125596.1 - 1 1366
AK289957.1 - 4 586
AK297013.1 - 5 425
BC001321.1 - 4 526 3456049
BM837218.1 - 4 619
CR457336.1 - 3 449

View Full Product Details