Gene Transcripts

Gene Symbol
CBWD5
Entrez Gene ID
220869
Gene Name
COBW domain containing 5
Gene Aliases
DC36
Chromosome Location
Chr. 9: 65666374 - 65734041 on Build GRCh38
UniGene ID
Hs.355950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286835.1 NP_001273764.1 12 1997
XM_005272748.1 XP_005272805.1 10 849
XM_005272750.1 XP_005272807.1 9 789
XM_005272751.1 XP_005272808.1 9 762
XM_006716914.1 XP_006716977.1 10 849
XM_017014455.1 XP_016869944.1 9 762
XM_017014456.1 XP_016869945.1 11 780
XM_017014457.1 XP_016869946.1 10 693
XM_017014459.1 XP_016869948.1 12 1028
XM_017014461.1 XP_016869950.1 11 941
GenBank mRNA AK097639.1 - 10 1382
AK292585.1 - 11 917
BC067803.1 - 12 1935 5294979
BC141856.1 - 11 915

Gene Symbol
CBWD1
Entrez Gene ID
55871
Gene Name
COBW domain containing 1
Gene Aliases
COBP
Chromosome Location
Chr. 9: 121038 - 179083 on Build GRCh38
UniGene ID
Hs.531330
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145355.1 NP_001138827.1 11 950
NM_018491.3 NP_060961.3 10 852
XM_005251513.4 XP_005251570.1 9 800
XM_005251514.1 XP_005251571.1 9 765
XM_011517962.2 XP_011516264.1 10 852
XM_011517963.1 XP_011516265.1 7 390
GenBank mRNA AF257330.2 - 10 810
AK290648.1 - 11 922
AK315855.1 - 10 776
AY343911.1 - 10 809
BC005996.1 - 10 852 4069589
BC009573.1 - 11 903 4133536
BC013432.1 - 10 823 4133722
BC132759.1 - 10 786 40146781

Gene Symbol
CBWD2
Entrez Gene ID
150472
Gene Name
COBW domain containing 2
Gene Aliases
Chromosome Location
Chr. 2: 113437691 - 113496204 on Build GRCh38
UniGene ID
Hs.531330
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172003.3 NP_742000.1 10 924
XM_005263602.2 XP_005263659.1 9 837
XM_005263603.2 XP_005263660.1 11 958
XM_011510667.1 XP_011508969.1 10 924
XM_011510668.2 XP_011508970.1 7 484
XM_011510669.2 XP_011508971.1 11 968
XM_017003408.1 XP_016858897.1 12 1088
GenBank mRNA AF452722.1 - 10 780
AK124867.1 - 1 2489
AK225251.1 - 11 938
AK290072.1 - 10 924
AY343910.1 - 10 809
BC120991.1 - 10 795 40116409
BC120992.1 - 10 795 40116411

Gene Symbol
CBWD3
Entrez Gene ID
445571
Gene Name
COBW domain containing 3
Gene Aliases
bA561O23.1
Chromosome Location
Chr. 9: 68232437 - 68300035 on Build GRCh38
UniGene ID
Hs.645337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291821.1 NP_001278750.1 9 773
NM_201453.3 NP_958861.2 10 833
XM_005277639.2 XP_005277696.1 9 825
XM_006717116.1 XP_006717179.1 10 912
XM_011518706.2 XP_011517008.1 5 363
XM_017014730.1 XP_016870219.1 8 765
XM_017014731.1 XP_016870220.1 10 3284
XM_017014732.1 XP_016870221.1 4 276
XM_017014733.1 XP_016870222.1 5 5583
GenBank mRNA AK297912.1 - 9 735
AY344633.1 - 10 809

Gene Symbol
CBWD6
Entrez Gene ID
644019
Gene Name
COBW domain containing 6
Gene Aliases
Chromosome Location
Chr. 9: 41131309 - 41198865 on Build GRCh38
UniGene ID
Hs.597638
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085457.1 NP_001078926.1 10 852
XM_017015019.1 XP_016870508.1 9 852
XM_017015020.1 XP_016870509.1 9 852
XM_017015022.1 XP_016870511.1 8 765
XM_017015024.1 XP_016870513.1 8 765
XM_017015026.1 XP_016870515.1 9 852
XM_017015027.1 XP_016870516.1 9 852
XM_017015028.1 XP_016870517.1 10 720
XM_017015029.1 XP_016870518.1 6 671
XM_017015031.1 XP_016870520.1 5 363
XM_017015033.1 XP_016870522.1 4 276
XM_017015034.1 XP_016870523.1 4 226
GenBank mRNA AF293368.1 - 3 157
AK293173.1 - 10 900
BC069006.1 - 11 993

View Full Product Details