Gene Transcripts

Gene Symbol
C4orf26
Entrez Gene ID
152816
Gene Name
chromosome 4 open reading frame 26
Gene Aliases
AI2A4
Chromosome Location
Chr. 4: 75554997 - 75565893 on Build GRCh38
UniGene ID
Hs.24510
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206981.1 NP_001193910.1 3 331
NM_178497.3 NP_848592.2 2 287
XM_011531668.2 XP_011529970.1 4 1286
GenBank mRNA AK074237.1 - 2 287
AK172776.1 - 2 287
AK300227.1 - 3 314
BC117342.1 - 2 287 40125893

View Full Product Details