Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290341.2 | NP_001277270.1 | 5 | 805 | |
NM_001290342.2 | NP_001277271.1 | 4 | 631 | ||
NM_001290343.1 | NP_001277272.1 | 4 | 542 | ||
NM_178842.4 | NP_849164.2 | 4 | 538 | ||
XM_011521355.2 | XP_011519657.1 | 5 | 848 | ||
XM_011521357.2 | XP_011519659.1 | 5 | 742 | ||
XM_011521358.1 | XP_011519660.1 | 5 | 626 | ||
XM_017022002.1 | XP_016877491.1 | 4 | 622 | ||
XM_017022003.1 | XP_016877492.1 | 4 | 547 | ||
XM_017022004.1 | XP_016877493.1 | 3 | 458 | ||
GenBank mRNA | AK307191.1 | - | 3 | 658 | |
BC028703.1 | - | 4 | 619 | 4837657 | |
BC034500.1 | - | 4 | 497 | 4838423 | |
BC034970.1 | - | 4 | 540 | 4822006 |