Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001031749.2 | NP_001026919.2 | 5 | 1540 | |
NM_001288763.1 | NP_001275692.1 | 4 | 1590 | ||
NM_182573.2 | NP_872379.2 | 5 | 1512 | ||
XM_011526794.2 | XP_011525096.1 | 5 | 1596 | ||
GenBank mRNA | AK055031.1 | - | 5 | 1490 | |
BC057816.1 | - | 5 | 1526 | 30345834 |