Gene Transcripts

Gene Symbol
KRT79
Entrez Gene ID
338785
Gene Name
keratin 79
Gene Aliases
K6L, KRT6L
Chromosome Location
Chr. 12: 52821447 - 52834293 on Build GRCh38
UniGene ID
Hs.711471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175834.2 NP_787028.1 2, 3 734
GenBank mRNA AJ564105.1 - 2, 3 719
BC039148.1 - 2, 3 736 4746574
BC063648.1 - 2, 3 733 4804408

View Full Product Details