Gene Transcripts

Gene Symbol
FGF4
Entrez Gene ID
2249
Gene Name
fibroblast growth factor 4
Gene Aliases
HBGF-4, HST, HST-1, HSTF1, K-FGF, KFGF
Chromosome Location
Chr. 11: 69773029 - 69775403 on Build GRCh38
UniGene ID
Hs.1755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002007.2 NP_001998.1 2, 3 770
GenBank mRNA AW340506.1 - 1, 2 102
M17446.1 - 2, 3 770

View Full Product Details