Gene Transcripts

Gene Symbol
TMPRSS9
Entrez Gene ID
360200
Gene Name
transmembrane protease, serine 9
Gene Aliases
Chromosome Location
Chr. 19: 2385254 - 2426261 on Build GRCh38
UniGene ID
Hs.465560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182973.1 NP_892018.1 11 1903
XM_011527978.2 XP_011526280.1 13 2266
XM_011527980.1 XP_011526282.1 5 922
XM_011527982.2 XP_011526284.1 3 697
GenBank mRNA AJ488946.1 - 11 1903
AJ488947.1 - 11 1903
AK131261.1 - 3 492

View Full Product Details