Gene Transcripts

Gene Symbol
IQCF5
Entrez Gene ID
389124
Gene Name
IQ motif containing F5
Gene Aliases
Chromosome Location
Chr. 3: 51873721 - 51875584 on Build GRCh38
UniGene ID
Hs.666604
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145059.1 NP_001138531.1 2 231
GenBank mRNA BX106781.1 - 2 231

View Full Product Details