Gene Transcripts

Gene Symbol
FLOT2
Entrez Gene ID
2319
Gene Name
flotillin 2
Gene Aliases
ECS-1, ECS1, ESA, ESA1, M17S1
Chromosome Location
Chr. 17: 28879339 - 28897728 on Build GRCh38
UniGene ID
Hs.514038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004475.2 NP_004466.2 10 1294
XM_005257950.3 XP_005258007.1 10 1325
XM_005257953.2 XP_005258010.1 9 1229
XM_017024394.1 XP_016879883.1 10 1222
XM_017024395.1 XP_016879884.1 11 1391
XM_017024396.1 XP_016879885.1 10 1228
GenBank mRNA AK225491.1 - 3 324
BC017292.1 - 9 1116
BT019477.1 - 8 1024
BT019478.1 - 8 1024
CR541784.1 - 8 1024
CR542265.1 - 8 1024
M60922.1 - 10 1150

View Full Product Details