Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000447.2 | NP_000438.2 | 7 | 947 | |
NM_012486.2 | NP_036618.2 | 7 | 947 | ||
XM_005273199.3 | XP_005273256.1 | 6 | 760 | ||
XM_017001835.1 | XP_016857324.1 | 6 | 1268 | ||
XM_017001836.1 | XP_016857325.1 | 6 | 760 | ||
GenBank mRNA | AF416718.1 | - | 5 | 588 | |
AK292299.1 | - | 8 | 899 | ||
AK301143.1 | - | 6 | 696 | ||
BC006365.2 | - | 8 | 1141 | 4131408 | |
BT006984.1 | - | 5 | 520 | ||
L43964.1 | - | 6 | 887 | ||
L44577.1 | - | 7 | 885 | ||
U34349.1 | - | 5 | 611 |