Gene Transcripts
- Gene Symbol
- C12orf57
- Entrez Gene ID
- 113246
- Gene Name
- chromosome 12 open reading frame 57
- Gene Aliases
- C10, GRCC10
- Chromosome Location
- Chr. 12: 6943438 - 6946003 on Build GRCh38
- UniGene ID
- Hs.405913
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BP375238.1 |
- |
2 |
384 |
|
Chr. 12 - 6944033 - 6945933 [+] |
GenBankmRNA |
AA371779.1 |
- |
2 |
285 |
|
Chr. 12 - 6944129 - 6944637 [+] |
View Full Product Details