Gene Transcripts

Gene Symbol
C15orf54
Entrez Gene ID
400360
Gene Name
chromosome 15 open reading frame 54
Gene Aliases
Chromosome Location
Chr. 15: 39250669 - 39254845 on Build GRCh38
UniGene ID
Hs.376109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302797.1 NP_001289726.1 2 375
NM_207445.3 NP_997328.1 2 486
GenBank mRNA AK096850.1 - 2 437
BC137300.1 - 2 389 9021297
BC137301.1 - 2 389 9021298
BC171789.1 - 2 279 9054443
BX647708.1 - 2 472

View Full Product Details