Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134.2 | NP_001125.1 | 8 | 976 | |
GenBank mRNA | AK297669.1 | - | 8 | 888 | |
AK297674.1 | - | 8 | 829 | ||
AK314817.1 | - | 8 | 970 | ||
BC027881.1 | - | 8 | 962 | 5208299 | |
V01514.1 | - | 8 | 976 |